To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of Differentiation 44 antigen
- Clonality:Monoclonal
- Clone:MEM-85
- Gene ID:960
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:CD44
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Antigen synonyms:MDU2|HUTCH-I|LHR|MC56|HCAM|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
- Storage buffer:Phosphate buffered saline pH 7.4 containing 15mM sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Tissue / cell preparation (Human). (Leucocytes of patient suffering from LGL type leukaemia).
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:>95% by SDS-PAGE
- Cat. no.:89361-180
- Supplier No.:GTX22212
Specifications
About this item
CD44 is a type 1 transmembrane glycoprotein also known as Phagocytic Glycoprotein-1 (pgp-1) and HCAM. CD44 is the receptor for hyaluronate and exists as a large number of different isoforms due to alternative RNA splicing. The major isoform expressed on lymphocytes, myeloid cells, and erythrocytes is a glycosylated type 1 transmembrane protein. Other isoforms contain glycosaminoglycans and are expressed on hematopoietic and non-hematopoietic cells. CD44 is involved in adhesion of leukocytes to endothelial cells, stromal cells, and the extracellular matrix.
ELISA: Use at an assay dependant concentration. FACS: Use at an assay dependant concentration. IP: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: MEM-85
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: