To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin Alpha 1, Skeletal Muscle
- Clonality:Polyclonal
- Gene ID:60
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Antigen symbol:ACTA1
- Conjugation:Unconjugated
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 and 0.01% (w/v) Sodium Azide
- Storage temperature:Store vial at –20 °C prior to opening. Aliquot contents and freeze at –20 °C or below for extended storage Avoid cycles of freezing and thawing.
- Concentration:1.3 mg/ml
- Immunogen:Synthetic peptide: KQEYDESGPSC conjugated to KLH, corresponding to amino acids 359-368 of Human beta Actin.
- Tested applications:WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89361-080
- Supplier No.:GTX21801
Specifications
About this item
The two major cytoskeletal proteins implicated in cell motility are actin and myosin. Actin and myosin are constituents of many cell types and are involved in a myriad of cellular processes including locomotion, secretion, cytoplasmic streaming, phagocytosis and cytokinesis. Although actin is one of the most conserved eukaryotic proteins, it is expressed in mammals and birds as at least six isoforms characterized by electrophoresis and amino acid sequence analysis. Four of them represent the differentiation markers of muscle tissues and two are found in practically all cells.
WB: Use at a dilution of 1:1000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: ACT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine