To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD34 molecule
- Clonality:Monoclonal
- Clone:4H11(APG)
- Gene ID:947
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CD34
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Size:100 μg
- Antigen synonyms:CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
- Storage buffer:Phosphate buffered saline pH 7.4 containing 15mM sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Tissue / cell preparation (Human). (Permanent human cell line derived from peripheral leucocytes of a patient suffering from chronic myeloid leukaemia).
- Tested applications:Blocking, Fluorescence-Activated Cell Sorting
- Purification:>95% by SDS-PAGE
- Cat. no.:89360-892
- Supplier No.:GTX20762
Specifications
About this item
CD34 is possibly a adhesion molecule with a putative role in early hematopoiesis by mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. It could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. CD34 is thought to have a role in presenting carbohydrate ligands to selectins. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction.
BL: Use at an assay dependant dilution. Completely blocks the binding of Class II monoclonal antibody QBEnd10 and Class III monoclonal antibodies BIRMA K3 and 8G12 in cross-blocking studies on a KG1a cell line. FACS: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: APG
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: