To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Raf-1 Proto-Oncogene, Serine/Threonine Kinase
- Clonality:Monoclonal
- Clone:410
- Gene ID:5894
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:RAF1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:Raf-1|CRAF|RAF|NS5|cRaf|C-raf|c-Raf|Proto-oncogene c-RAF
- Storage buffer:Lyophilized from a solution in 0.01M PBS pH7.2
- Molecular weight:75 kDa
- Storage temperature:Upon receipt - Keep as concentrated solution. Aliquot and store at -20oC or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Purified recombinant human Raf-1 (mw:75 kDa).
- Tested applications:ELISA, WB
- Purification:n/a
- Cat. no.:89360-856
- Supplier No.:GTX20656
Specifications
About this item
The Raf kinases are important inter-mediates in signal transduction. Raf-l proteins have intrinsic serine/threonine kinase activity. Interaction between Ras proteins and Raf proteins results in Raf-mediated phosphorylation and activation of MEK (also known as MAP kinase kinase).
For WB: A concentration of 1.0 ?g/ml, will allow visualization of 0.2 ?g/lane of human Raf-1. For ELISA: A concentration of 1.0 ?g/ml, will allow visualization of 0.2 ?g/lane of human Raf-1. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user
Type: Primary
Antigen: RAF1
Clonality: Monoclonal
Clone: 410
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human