Order Entry
ContactUsLinkComponent
Anti-CD44 Rat Monoclonal Antibody [clone: IM7]
Anti-CD44 Rat Monoclonal Antibody [clone: IM7]
Catalog #: 89351-490
Supplier:  Genetex
Anti-CD44 Rat Monoclonal Antibody [clone: IM7]
Catalog #: 89351-490
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cluster of Differentiation 44 antigen
  • Clonality:
    Monoclonal
  • Clone:
    IM7
  • Gene ID:
    960
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Cat
  • Antigen symbol:
    CD44
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    125 μg
  • Antigen synonyms:
    MDU2|HUTCH-I|LHR|MC56|HCAM|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
  • Storage buffer:
    Preservative: 0.1% Sodium Azide. Constituents: 1.0% BSA, 0.01M PBS, pH 7.2
  • Storage temperature:
    Store at 4°C. Do not freeze.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Unfortunately, this information is considered to be commercially sensitive
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Protein A purified
  • Cat. no.:
    89351-490
  • Supplier No.:
    GTX19629

Specifications

About this item

CD44 is a type 1 transmembrane glycoprotein also known as Phagocytic Glycoprotein 1 (pgp 1) and HCAM. CD44 is the receptor for hyaluronate and exists as a large number of different isoforms due to alternative RNA splicing. The major isoform expressed on lymphocytes, myeloid cells, and erythrocytes is a glycosylated type 1 transmembrane protein. Other isoforms contain glycosaminoglycans and are expressed on hematopoietic and non hematopoietic cells. CD44 is involved in adhesion of leukocytes to endothelial cells, stromal cells, and the extracellular matrix.

FACS: Use 1?g for 10^6 cells. IHC-Fr (acetone fixed): Use at an assay dependent dilution. IP: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:

Customers Who Bought This Also Bought