To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Klotho
- Clonality:Polyclonal
- Gene ID:16591
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:KL
- Conjugation:Unconjugated
- ELISA:Yes
- Size:25μg
- Western blot:Yes
- Storage buffer:100 mM Tris-HCl, 0.2% BSA, pH 7.5, with 0.05% sodium azide as preservative
- Storage temperature:Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide of 17 amino acids (Mouse) (C terminal).
- Tested applications:ELISA, WB
- Purification:Affinity purified with antigen
- Cat. no.:89360-408
- Supplier No.:GTX18131
Specifications
About this item
Animal models of human aging which display the characteristic diseases associated with aging provide insight into the cause of such diseases. Recently a transgenic mouse model, called Klotho, with several premature aging phenotypes has been described. They display premature aging phenotypes such as Osteoporosis, age related skin changes, ectopic calcifications, atrophy of genital organs and thymus, emphysema and short life span. KL protein associated with the Klotho mutation is a 1014 amino acid long peptide. It has a putative signal sequence at its N-terminus and a single transmembrane domain near its C-terminus, which is postulated to anchor it to the membrane. kl gene expression was observed to be tissue specific. Improvement of systemic aging phenotypes in kl/kl mice occurs even when the exogenous expression was limited to some organs, suggesting that KL associated aging is regulated through a humoral signaling pathway. KL has been reported to be localized on the cell surface when expressed on Cho cells. Human kl cDNA is expected to encode a protein of 1012 amino acids.
For ELISA: Use at a dilution of 1:10,000 - 1:100,000. For WB: Use at a concentration of 1 - 10 ?g/ml (chemiluminescence). Predicted molecular weight: 116 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: KL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse