To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Synaptobrevin
- Clonality:Monoclonal
- Clone:4E240
- Gene ID:6843
- Host:Mouse
- Isotype:IgM
- Reactivity:Bovine
- Antigen symbol:VAMP1
- Conjugation:Unconjugated
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:SYB1|SPAX1|VAMP-1
- Storage buffer:Preservative: None. Constituents: 50% Glycerol, PBS, 0.1mM PMSF. pH 7.2
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Tissue / cell preparation (Human).Mice were immunized with synaptic vesicle containing fractions which were immunoprecipitated from human brain homogenates using anti-human synaptophysin monoclonal antibodies.
- Tested applications:WB
- Purification:Protein G purified
- Cat. no.:89360-372
- Supplier No.:GTX18013
Specifications
About this item
Synaptobrevins (VAMP1), syntaxins, and the 25 kDa synaptosomal associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. VAMP1 is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family.
WB: 1/1000. Predicted molecular weight: 13 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: VAMP1
Clonality: Monoclonal
Clone: 4E240
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Monkey, Pig, Rat, Guinea Pig, Hamster, Rabbit, Sheep, Xenopus laevis, Bovine