To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 6
- Clonality:Polyclonal
- Gene ID:3569
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:IL6
- Conjugation:Biotin
- ELISA:Yes
- Size:25μg
- Western blot:Yes
- Antigen synonyms:HSF|HGF|IFNB2|interferon beta 2|BSF2|IL-6
- Storage buffer:Phosphate-buffered saline, pH 7.3 containing 0.1% BSA
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.05 mg/ml
- Immunogen:Recombinant full length protein (Human).
- Tested applications:ELISA, WB
- Purification:Immunogen affinity purified
- Cat. no.:89360-314
- Supplier No.:GTX17529
Specifications
About this item
Interleukin 6 (IL6) is a multifunctional 24 kD protein originally discovered in the medium of RNA stimulated fibroblastoid cells. It is upregulated by IL1, TNF, PDGF, IFN beta, TNF alpha, NGF, IL17 and downregulated by glucocorticoids IL4, TGF beta. IL6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be as important as IL1 and TNF alpha in regulating the acute phase response. IL6 has also been implicated in regulating adipose mass. IL6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. It acts upon a variety of cells including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. In addition, IL6 appears to interact with IL2 in the proliferation of T lymphocytes. IL6 potentiates the proliferative effect of IL3 on multipotential hematopoietic progenitors.
ELISA: Use at a concentration of 0.15 - 0.3 ?g/ml. This concentration allows the detection of at least 0.2 ng/well of recombinant hIL6. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. The detection limit for recombinant hIL6 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions. Predicted molecular weight: 26 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human