Order Entry
ContactUsLinkComponent
Anti-CD15 Mouse Monoclonal Antibody [clone: MMA]
Anti-CD15 Mouse Monoclonal Antibody [clone: MMA]
Catalog #: 89351-314
Supplier:  Genetex
Mouse Monoclonal antibody to CD15 (fucosyltransferase 4)
Anti-CD15 Mouse Monoclonal Antibody [clone: MMA]
Catalog #: 89351-314
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    3-fucosyl-N-acetyl-lactosamine
  • Clonality:
    Monoclonal
  • Clone:
    MMA
  • Gene ID:
    14345
  • Host:
    Mouse
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Antigen symbol:
    CD15
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    1 ml
  • Antigen synonyms:
    CD15|Granulocyte Associated Antigen|MC-480|BG-7|SSEA-1|SIG-3339-1000
  • Storage buffer:
    Preservative: Sodium Azide.
  • Storage temperature:
    Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
  • Immunogen:
    BALB/C mice were injected with U937 histiocytic cell line.
  • Tested applications:
    IHC-P
  • Purification:
    Purified IgM
  • Cat. no.:
    89351-314
  • Supplier No.:
    GTX17080

Specifications

About this item

The CD15 antigen is expressed on approximately 90% human circulating granulocytes (membranes and granules), 30 to 60% of circulating monocytes and is absent from normal lymphocytes. CD15 antigen is also expressed on Reed Sternberg cells of Hodgkin's disease, on T cell lymphomas including mycosis fungoides and on some leukemias. Outside the hematopoietic system, CD15 expression is described in certain normal and neoplastic epithelial cells and in astrocytes. CD15 antibodies recognize the tri saccharide 3 fucosyllactosamine (3Fl) which is present in lacto N fucopentaose III and in the blood group antigen X hapten. At least 5 major CD15 antigens (105, 135, 165, 185, 220 kDa) are present on the surface membranes of polymorphonuclear cells. The hapten occurs also in glycolipids. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis.

IHC-P: 1/25 - 1/50. Perform heat mediated antigen retrieval before commencing with IHC staining protocol. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Customers Who Bought This Also Bought