Order Entry
ContactUsLinkComponent
Anti-CAPN2 Mouse Monoclonal Antibody [clone: 28F3]
Anti-CAPN2 Mouse Monoclonal Antibody [clone: 28F3]
Catalog #: 89360-252
Supplier:  Genetex
Anti-CAPN2 Mouse Monoclonal Antibody [clone: 28F3]
Catalog #: 89360-252
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calpain 2
  • Clonality:
    Monoclonal
  • Clone:
    28F3
  • Gene ID:
    824
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Bovine
  • Antigen symbol:
    CAPN2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    100 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    CANP2|CANPL2|mCANP|FLJ39928|Millimolar-calpain|EC 3.4.22.53|M-calpain|CANPml
  • Storage buffer:
    Preservative: 3mM Sodium Azide. Constituents: 50% Glycerol, 0.15M Sodium chloride, 0.02M Sodium phosphate. pH 7.5.
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Full length protein (Human). Antibody specifically recognizes small subunit.
  • Tested applications:
    ELISA, IP, WB
  • Purification:
    Protein G purified
  • Cat. no.:
    89360-252

Specifications

About this item

Mu Calpain, and m calpain, also known as Calpain 2, are intracellular, calcium dependent cysteine proteases. Mu calpain has a micromolar sensitivity (thus the mu) as compared to the millimolar calcium sensitivity of m calpain. Both Calpains 1 and 2 are composed of an 80 kD subunit and a 30 kD subunit. Whereas the 30 kDa subunit is shared by both enzymes, the larger catalytic subunits are different and exhibit the distinct Ca++ requirements that are suggested by their names. The calpains have papain like activity, thus the pain nomenclature. Both Calpain 1 and Calpain 2 are ubiquitously expressed, and are countered by the endogenous calpain inhibitor, calpastatin.Other calpain family members (calpain 94, ncl2, ncl3, etc) have more limited tissue distribution, and perhaps different functions. The calpain family members consist of a common small subunit (Calpain 4), and a large variable subunit. It is not clear that all calpains contain a small subunit. Domains in the large subunit include the amino terminal domain I, the proteinase domain II, domain III, and the EF hand domain IV. The calpains appear to serve multiple physiological roles, and ideas concerning the functions of these enzymes are in a state of rapid flux.

ELISA: Use at a concentration of 0.5 - 1 ?g/ml. IP: Use at a concentration of 1 - 2 ?g/ml. WB: Use at a concentration of 0.5 - 1 ?g/ml. Predicted molecular weight: 30 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CAPN2
Clonality: Monoclonal
Clone: 28F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine

Customers Who Bought This Also Bought