To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Vascular Endothelial Growth Factor
- Clonality:Monoclonal
- Clone:14-124
- Gene ID:7422
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:VEGF
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:500 μl
- Western blot:Yes
- Antigen synonyms:VEGFA|VEGF-A|Vascular Permeability Factor|VPF|Vascular Endothelial Growth Factor
- Storage buffer:50 mM sodium phosphate buffer, 0.2% gelatin, pH 7.5 with =0.1% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.1 mg/ml
- Immunogen:Synthetic peptide (Human) (N terminal) of VEGF165.
- Tested applications:IHC-P
- Purification:Purified IgG
- Cat. no.:89351-270
- Supplier No.:GTX16883
Specifications
About this item
VEGF (Vascular Endothelial Growth Factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF 165 and VEGF 121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF 189 and VEGF 206) remain cell associated.
For IHC-P: Use at a concentration of 4 - 6?g/ml. For WB: Use at a concentration of 0.5 - 1?g/ml. Predicted molecular weight: 46 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: VEGF
Clonality: Monoclonal
Clone: 14-124
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human