Order Entry
ContactUsLinkComponent
Anti-CD44 Mouse Monoclonal Antibody [clone: 156-3C11]
Anti-CD44 Mouse Monoclonal Antibody [clone: 156-3C11]
Catalog #: 89351-240
Supplier:  Genetex
Mouse Monoclonal antibody to CD44 (CD44 molecule (Indian blood group))
Anti-CD44 Mouse Monoclonal Antibody [clone: 156-3C11]
Catalog #: 89351-240
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cluster of Differentiation 44 antigen
  • Clonality:
    Monoclonal
  • Clone:
    156-3C11
  • Gene ID:
    960
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Antigen symbol:
    CD44
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    500 μl
  • Antigen synonyms:
    MDU2|HUTCH-I|LHR|MC56|HCAM|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
  • Storage buffer:
    Contains sodium azide
  • Storage temperature:
    Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
  • Immunogen:
    Stimulated human leukocytes
  • Tested applications:
    IHC-P
  • Purification:
    Protein A purified
  • Cat. no.:
    89351-240
  • Supplier No.:
    GTX16728

Specifications

About this item

This antibody is specific to a cell surface glycoprotein of 80-95 kD, known as CD44. CD44 is expressed on the surface of lymphocytes, monocytes and granulocytes. This antibody reacts with red cells pretreated with 2-aminoethylisothiouroniumbromide (AET).

For IHC-P: Use at a dilution of 1:25-1:75 in an ABC method. We suggest an incubation period of 30 minutes at RT. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 156-3C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Customers Who Bought This Also Bought