To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Lysophosphatidic acid receptor 3
- Clonality:Polyclonal
- Gene ID:23566
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:LPA3
- Conjugation:Unconjugated
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:HOFNH30|RP4-678I3|LPAR3|GPCR|EDG7|LPA3|Edg-7|LP-A3
- Storage buffer:PBS, pH7.2 containing 0.08% sodium azide as preservative
- Storage temperature:Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide - C terminal of Human EDG7 - Immunogen sequence information is commercially sensitive
- Tested applications:WB
- Purification:IgG fraction
- Cat. no.:89360-214
- Supplier No.:GTX16456
Specifications
About this item
EDG7 is a member of the G protein-coupled receptor family, as well as the EDG family of proteins. This protein functions as a cellular receptor for lysophosphatidic acid and mediates lysophosphatidic acid-evoked calcium mobilization. This receptor couples predominantly to G(q/11) alpha proteins.EDG7 has been reported in human prostate, as well as rodent brain, kidney, lung, placenta, prostate, and testis. ESTs have been isolated from a normal prostate library, as well as cancer libraries of the head/neck, ovary, and thyroid.
For WB: Use at an assay dependent dilution. Predicted molecular weight: 42.36 kDa. Due to low expression of EDG receptors, we recommend use of Pierce Femto Signal substrate for western blot development. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: LPA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human