To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Transforming Growth Factor Beta 3
- Clonality:Polyclonal
- Gene ID:7043
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TGFB3
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:500 μl
- Western blot:Yes
- Antigen synonyms:RNHF|ARVD|ARVD1|TGF-beta3
- Storage buffer:Phosphate-buffered saline, pH 7.4 containing 1% BSA and 0.09% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:A synthetic peptide derived from the C-terminus of the precursor form of human TGF beta 3.
- Tested applications:IHC-P, IP, WB
- Purification:Immunogen affinity purified
- Cat. no.:89351-162
- Supplier No.:GTX15537
Specifications
About this item
Transforming Growth Factor beta 3 (TGF beta 3) is a member of the TGF beta family of growth factors. The TGF beta polypeptides are multifunctional, capable of influencing cell proliferation, differentiation, and other functions in a wide range of cell types. The multi-modal nature of TGF beta is seen in its ability to stimulate or inhibit cellular proliferation. In general, cells of mesenchymal origin appear to be stimulated by TGF beta whereas cells of epithelial or neuroectodermal origin are inhibited by the peptide.
IHC-P: Use at a dilution of 1/100 for 10 minutes at RT. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10 minutes followed by cooling at RT for 20 minutes. GTX15537 demonstrates cytoplasmic staining. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 47 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: TGFB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat