To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Phospholamban
- Clonality:Polyclonal
- Gene ID:5350
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Antigen symbol:PLN
- Conjugation:Unconjugated
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:phospholamban|PLN|CMD1P|PLB
- Modification:phospho-Ser16
- Storage buffer:Preservative: 0.05% Sodium Azide; Constituents: 30% Glycerol, 0.2M Tris glycine, 0.15M Sodium chloride
- Storage temperature:Stable for 2 years at -20 degrees Celcius from the date of shipment.
- Concentration:1 mg/ml
- Immunogen:KLH-conjugated, synthetic peptide corresponding to amino acids 14-25 of human cardiac phospholamban (RA[pS]TIEMPQQAR-C) with an added C-terminal cysteine. The immunizing sequence is identical in mouse, rat, rabbit, pig and dog.
- Tested applications:WB
- Purification:Immunogen affinity purified
- Cat. no.:89360-082
- Supplier No.:GTX15000
Specifications
About this item
Phospholamban (PLB) is a 52 amino acid phosphoprotein which regulates the calcium pump of cardiac sarcoplasmic reticulum (SR). PLB is an oligomer of five identical subunits each having a cytoplasmic and transmembrane domain. The cytoplasmic domain (residues 1 to 25) contains the phosphorylation sites and is highly basic and readily cleaved by proteases; whereas the transmembrane domain (residues 25 to 52) is mostly hydrophobic, protease resistant and stabilizes the pentamer.
WB: Use at a concentration of 0.5 - 2 ?g/ml. Predicted molecular weight: 5-25 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: PLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine