To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C Motif) Receptor 2
- Clonality:Polyclonal
- Gene ID:3579
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CXCR2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:IL8RB|CD182|IL8R2|CDw128b|IL8RA|mIL-8RH|Il8rb|CMKAR2|IL-8Rh|IL-8rb
- Storage buffer:Preservative: 0.02% Sodium Azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Synthetic peptide (selective for CXCR2 receptor +C-terminal region)
- Tested applications:IHC-Fr, IP, WB
- Purification:Immunogen affinity purified
- Cat. no.:89360-070
- Supplier No.:GTX14935
Specifications
About this item
CXCR2 is a receptor for interleukin 8, which is a powerful neutrophil chemotactic factor. It is a member of the GPCR family (subfamily, chemokine). Binding of IL8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL8 with a high affinity and to GRO/MGSA and NAP2 also with a high affinity. It has been reported to be expressed in a wide variety of tissues. ESTs have been isolated from human placenta and thymus libraries.
IHC: Use at an assay dependent dilution. IP: Use at a dilution of 1/250. WB: Use at a dilution of 1/500. Predicted molecular weight: 55 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CXCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: