To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Phosphodiesterase 6G
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:PDE6G
- Conjugation:Unconjugated
- Western blot:Yes
- Storage buffer:Preservative: 0.02% Sodium Azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) (C terminal).
- Tested applications:WB
- Purification:Immunogen affinity purified
- Cat. no.:89367-368
- Supplier No.:GTX14627
Specifications
About this item
Cyclic GMP-phosphodiesterase plays a key role in the normal functioning of retinal rod photoreceptor cells. The enzyme is composed of alpha- and beta-catalytic subunits which are inhibited by 2 identical gamma subunits. The PDE6 gamma subunit has been cloned and sequenced from human retina. The 1,012-bp cDNA had a coding region of 261 bp that was highly homologous to those of the corresponding cDNA from bovine and mouse retinas. Comparison of the deduced amino acid sequences from 3 species indicated evolutionary conservation.
Type: Primary
Antigen: PDE6G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human