To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Complement Component 1, Q Subcomponent, A Chain
- Clonality:Polyclonal
- Gene ID:712
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Antigen symbol:C1Q
- Conjugation:Unconjugated
- ELISA:Yes
- Size:50 μg
- Western blot:Yes
- Storage buffer:Phosphate-buffered saline, no preservative added
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Full length plasma C1q protein (Human).
- Tested applications:ELISA, WB
- Purification:Affinity purified with antigen
- Cat. no.:89367-222
- Supplier No.:GTX14004
Specifications
About this item
C1q is a sub-component of the first complement component C1, which is formed by the interaction of C1q with the tetramer C1s-C1r-C1r-C1s.
For ELISA: Use at a dilution of 0.05 - 0.1 ?g/ml. For WB: Use at an assay dependent dilution. Predicted molecular weight: 26 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: C1Q
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human