Order Entry
ContactUsLinkComponent
Anti-CENPA Mouse Monoclonal Antibody [clone: 42448]
Anti-CENPA Mouse Monoclonal Antibody [clone: 42448]
Catalog #: 89359-914
Supplier:  Genetex
Mouse Monoclonal antibody to CENPA (centromere protein A)
Anti-CENPA Mouse Monoclonal Antibody [clone: 42448]
Catalog #: 89359-914
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Centromere Protein A
  • Clonality:
    Monoclonal
  • Clone:
    3-19
  • Gene ID:
    1058
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Neutralisation:
    Yes
  • Reactivity:
    Human
  • Antigen symbol:
    CENPA
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    50 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    centromeric protein A|Histone H3 like centromeric protein A.|CENP-A|centromere protein A|Centromere autoantigen A|CENP A|Centromere protein A 17kDa
  • Amino acid number:
    3 to 19
  • Storage buffer:
    Preservative: None Constituents: 50% Glycerol, PBS. pH 7.2
  • Sequence:
    PRRRSRKPEAPRRRSPS
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide: PRRRSRKPEAPRRRSPS, corresponding to amino acids 3-19 of Human CENP A.
  • Tested applications:
    ICC/IF, IHC, Neut, WB
  • Purification:
    Protein A purified
  • Cat. no.:
    89359-914
  • Supplier No.:
    GTX13939

Specifications

About this item

Centromeres are the differentiated chromosomal domains that specify the mitotic behavior of chromosomes. CENPA encodes a centromere protein which contains a histone H3 related histone fold domain that is required for targeting to the centromere. CENPA is proposed to be a component of a modified nucleosome or nucleosome-like structure in which it replaces 1 or both copies of conventional histone H3 in the (H3-H4)2 tetrameric core of the nucleosome particle.

ICC: Use at a concentration of 10 ?g/ml. IHC: Use at a concentration of 5 ?g/ml. For paraffin embedded sections, heat treatment may be necessary. Microwave sections 2 times for 10 minutes each in citrate buffer (pH 6.5). WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately 18 kDa (predicted molecular weight: 16 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user. User review: This antibody is great for immunofluorescence on human cells (primary fibroblasts and transformed cell lines such as HeLa, HT1080, A549, H1299). Works well with various fixation conditions, including 1-4% paraformaldehyde (PFA), -20C methanol, 2% PFA followed by -20C methanol, and on unfixed preparations. We routinely use it at a dilution of 1:200-1:400.

Type: Primary
Antigen: CENPA
Clonality: Monoclonal
Clone: 42448
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Customers Who Bought This Also Bought