Order Entry
ContactUsLinkComponent
Anti-RPE65 Mouse Monoclonal Antibody
Anti-RPE65 Mouse Monoclonal Antibody
Catalog #: 89359-864
Supplier:  Genetex
Anti-RPE65 Mouse Monoclonal Antibody
Catalog #: 89359-864
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Retinal Pigment Epithelium-specific 65 kDa protein
  • Clonality:
    Monoclonal
  • Gene ID:
    282043
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    RPE65
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    100 μl
  • Western blot:
    Yes
  • Antigen synonyms:
    sRPE65|all-trans-retinyl-palmitate hydrolase|retinol isomerase|retinoid isomerohydrolase|retinitis pigmentosa 20 (autosomal recessive)|retinal pigment epithelium-specific 65 kDa protein|RBP-binding membrane protein|p63|LCA2|RP20|rd12|mRPE65
  • Storage buffer:
    Preservative: 0.1% Sodium Azide; Constituents: PBS
  • Storage temperature:
    Store at –20 degrees C. Avoid repeated freeze-thaw cycles.
  • Concentration:
    0.96 mg/ml
  • Immunogen:
    Bovine RPE microsomal membrane proteins.
  • Tested applications:
    IHC, WB
  • Purification:
    Protein A purified
  • Cat. no.:
    89359-864
  • Supplier No.:
    GTX13826

Specifications

About this item

Highly conserved among vertebrate species, RPE65 is a major protein of the retinal pigment epithelium (RPE). RPE65 is essential for the regeneration of rhodopsin in the visual cycle. Mutations in RPE65 are responsible for certain forms of autosomal recessive severe retinal dystrophy, including Leber Congenital Amaurosis (LCA). Loss of function results in a block in retinoid processing required for the synthesis of 11-cis retinal and the accumulation of retinyl esters in the RPE.

By Western blot, this antibody recognizes a band at ~65 kDa, representing bovine RPE65 protein. It has also been tested on recombinant human protein in COS7 cells. It may also be used in immunohistochemistry on murine retinal sections and ICC in porcine.Suggested working dilutions: *Western Blot - 1:5,000-1:10,000 (ECL or Alk. Phos.). xpression in primary culture of RPE cells is undetectable in Western blot after day 14. Established cell lines are similarly devoid of RPE65 protein, as indicated in western blot. However, mRNA levels are detectable by Northern for at least 7 weeks. Immunohistochemistry - 1:250 (frozen sections)*The investigator should determine the optimal working dilution for a specific application.

Type: Primary
Antigen: RPE65
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Customers Who Bought This Also Bought