To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Superoxide dismutase 2, mitochondrial
- Clonality:Polyclonal
- Gene ID:6648
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Antigen symbol:SOD2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:mangano-superoxide dismutase|MVCD6|manganese-containing superoxide dismutase|IPO-B|indophenoloxidase B|Mn-SOD|IPOB|superoxide dismutase|MNSOD|manganese superoxide dismutase|Mn superoxide dismutase
- Storage buffer:50% Glycerol, 50mM Sodium Phosphate. pH 7.2 containing 0.1%BSA and 0.1% Sodium Azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Full length protein (Human).
- Tested applications:Enzyme Immunoassay
- Purification:Antigen affinity purification
- Cat. no.:89359-732
- Supplier No.:GTX13533
Specifications
About this item
Superoxide dismutase (SOD) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells. They are: extracellular SOD (EC SOD), copper and zinc-containing SOD (Cu/Zn SOD) and manganese-containing SOD (Mn SOD). The Cu/Zn form contains Cu and Zn ions and exists as a 32 kDa dimer in the cytosol. Mn SOD is an 80 kDa tetramer that contains Mn ion and resides in the mitochondrial matrix. Mn SOD is a tumor necrosis factor (TNF)-inducible enzyme that protects cells from TNF-mediated apoptosis via superoxide anion detoxification and the subsequent regulation of apoptosis through cytochrome c release and the modulation of the redox state of the mitochondria. Mn SOD has also been shown to be a tumor suppressor in human breast cancer. Overexpression of this enzyme protects neurons from NMDA- and nitric oxide-induced neurotoxicity.
EIA: Use at an assay dependent dilution. IHC: Use at a concentration of 1 ?g/ml. IP: Use at a concentration of 10 ?g/ml. WB: Use at a dilution of 1/5000. Detects a band of approximately 25 kDa (predicted molecular weight: 26.6 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine