Order Entry
ContactUsLinkComponent
Anti-HSP90 Mouse Monoclonal Antibody [clone: AC88]
Anti-HSP90 Mouse Monoclonal Antibody [clone: AC88]
Catalog #: 89359-690
Supplier:  Genetex
Anti-HSP90 Mouse Monoclonal Antibody [clone: AC88]
Catalog #: 89359-690
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Monoclonal
  • Clone:
    AC88
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Carp
  • Antigen symbol:
    HSP90
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    100 μg
  • Western blot:
    Yes
  • Storage buffer:
    Preservative: NoneConstituents: 50% Glycerol, PBS, 0.1mM PMSF. pH 7.2
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Achlya ambisexualis (water mold) Hsp90.
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Protein G purified
  • Cat. no.:
    89359-690
  • Supplier No.:
    GTX13492

Specifications

About this item

The 90kDa molecular chaperone family comprises several proteins including the 90kDa heat shock protein, Hsp90 and the 94kDa glucose regulated protein, grp94 which are major molecular chaperones of the cytosol and of the endoplasmic reticulum. In mammalian cells there are at least two Hsp90 isoforms, Hsp90a and hsp90s which are encoded by separate genes. The amino acid sequence of human and yeast Hsp90a is 85% and 90% homologous to that of Hsp90s respectively. All known members of the Hsp90 protein family are highly conserved, especially in the N terminal and C terminal regions which have been shown to contain independent chaperone sites with different substrate specificity. These ubiquitous and highly conserved proteins account for 1-2% of all cellular proteins in most cells. Hsp90 is part of the cell's powerful network of chaperones to fight the deleterious consequences of protein unfolding caused by nonphysiological conditions. However, in the absence of stress, Hsp90 is a necessary component of fundamental cellular processes such as hormone signaling and cell cycle control. In this context several key regulatory proteins such as steriod receptors, cell cycle kinases involved in signal transduction and p53 have been identified as substrates of Hsp90. It has been suggested that Hsp90 acts as a capacitor for morphological evolution by buffering widespread variation, which may affect morphogenic pathways.

IHC-P: Use at an assay dependent dilution.When used in immunoprecipitation, this antibody does not appear to react well with Hsp90 that is bound to steroid receptors or to pp60v-src of Rouse sarcoma virus. WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately 90 kDa (predicted molecular weight: 84.7 kDa(alpha) and 83.2 kDa (beta)). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HSP90
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Carp

Customers Who Bought This Also Bought