Order Entry
ContactUsLinkComponent
Anti-AURKA Rabbit Polyclonal Antibody
Anti-AURKA Rabbit Polyclonal Antibody
Catalog #: 89359-668
Supplier:  Genetex
Anti-AURKA Rabbit Polyclonal Antibody
Catalog #: 89359-668
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Aurora 2
  • Clonality:
    Polyclonal
  • Gene ID:
    6790
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    AURKA
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    50 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    BTAK|AURA|STK7|ARK1|PPP1R47|Serine/threonine-protein kinase 12|STK6|AURORA2|Aurora-B|STK15|AIK
  • Storage buffer:
    Preservative: 0.1% Sodium Azide; Constituents: Tris-citrate/phosphate. pH 7-8
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide exons 5 and 6 (Human).
  • Tested applications:
    ICC/IF, IP, WB
  • Purification:
    Purified IgG
  • Cat. no.:
    89359-668
  • Supplier No.:
    GTX13408

Specifications

About this item

Aurora A may play a role in cell cycle regulation during anaphase and/or telophase, in relation to the function of the centrosome/spindle pole region during chromosome segregation. It maybe involved in microtubule formation and/or stabilization and may also play a key role during tumor development and progression.

IF: Use at a concentration of 1 ?g/ml. IP: Use at a concentration of 1 - 5 ?g/mg lysate. WB: Use at a concentration of 0.05 - 0.2 ?g/ml. Predicted molecular weight: 46 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: AURKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought