To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-5
- Clonality:Polyclonal
- Gene ID:5868
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Antigen symbol:RAB5
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:RAB5A|Rab5a|RAB 5|RAS|RAB 5A|RAB5|RAS associated protein
- Storage buffer:Preservative: None Constituents: Whole Serum
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide: PKNEPQNPGANSARGR conjugated to KLH, corresponding to amino acids 182-197 of Human Rab5.
- Tested applications:ICC/IF, IHC, IP, WB
- Purification:Whole antiserum
- Cat. no.:89359-618
- Supplier No.:GTX13253
Specifications
About this item
Rab5 can be used as a marker for early endosomes. See Clague et al. 1998 for more information.
ICC: Use at an assay dependent dilution. IHC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/2000. Detects a band of approximately 26 kDa (predicted molecular weight: 25.8 kDa). An additional higher molecular mass band of unknown identity at ~45 kDa is detected in some samples on immunoblots. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: RAB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine