To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Brain Natriuretic Peptide
- Clonality:Monoclonal
- Clone:15F11
- Gene ID:4879
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:NPPB
- Conjugation:Unconjugated
- ELISA:Yes
- Size:200μg
- Western blot:Yes
- Antigen synonyms:BNP|natriuretic peptide B
- Storage buffer:Phosphate buffered, pH 7.4, saline containing 0.1% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:5 mg/ml
- Immunogen:Synthetic peptide: ETSGLQEQRNHLQGK, corresponding to amino acids 13 - 27 of Human proBNP.
- Tested applications:ELISA, WB
- Purification:Protein A purified
- Cat. no.:89350-870
- Supplier No.:GTX13115
Specifications
About this item
In cardiac tissue brain natriuretic peptide (BNP) is synthesized as 134 amino acid precursor (prepro-BNP), which is cleaved by proteases to form a 26 aa ""signal"" peptide and a 108 aa pro-BNP. Proteolytic digestion of pro-BNP results in formation of 76 aa amino-terminal NT-proBNP and biologically active 32 aa BNP hormone molecule. Both proBNP and NTproBNP circulate in human plasma and have been proposed as markers for early diagnosis of left ventricular dysfunction as well as prognostic markers of possible cardiac complications at patients with heart failure.
For ELISA: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Can be paired for ELISA with Mouse monoclonal [24E11] to proBNP (GTX13123). Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: NPPB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human