To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:MAS1 oncogene
- Clonality:Polyclonal
- Gene ID:4142
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:MAS1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:25μg
- Antigen synonyms:MGC119966|MGRA|MAS
- Storage buffer:Phosphate-buffered saline, pH 7.7, containing 0.01% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at -70 °C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive)
- Tested applications:IHC-P
- Purification:Immunogen affinity purified
- Cat. no.:89359-556
- Supplier No.:GTX13032
Specifications
About this item
The structure of the MAS1 product indicates that it belongs to the class of receptors that are coupled to GTP-binding proteins and share a conserved structural motif, which is described as a '7-transmembrane segment' following the prediction that these hydrophobic segments form membrane-spanning alpha-helices. The MAS1 protein may be a receptor that, when activated, modulates a critical component in a growth-regulating pathway to bring about oncogenic effects.
For IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human