To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Heat Shock Protein 25
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Guinea pig
- Antigen symbol:HSP25
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:Stress-responsive protein 27|Heat shock 27 kDa protein|SRP27|HSP27|HspB1|HSP25|Estrogen-regulated 24 kDa protein|HSPB1|HSP 27|Heat shock protein beta-1|28 kDa heat shock protein
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant mouse Hsp25
- Tested applications:IP, WB
- Purification:Protein A purified
- Cat. no.:89359-340
Specifications
About this item
In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock or stress proteins. One class of stress proteins, termed the ""small heat shock proteins"" is comprised of a diverse group of proteins from ~15 to >30kDa, with a single form found in yeast and human cells and multiple related forms found in higher plants. A direct relationship between elevated stress protein expression and neoplasia has been suggested from studies on estrogen-induced proteins in breast cancer cells. In addition, Hsp27 exhibits rapid increased phosphorylation in response to various mitogens, tumor promoters (e.g. phorbol esters) and calcium ionophores. The expression of a 24kDa protein was shown to correlate with steroid hormone receptors in cell lines, organs , and tumors. The 24kDa estrogen-induced protein was found to be expressed at high levels in carcinoma of the breast. Another study has suggested that the levels of Hsp27 correlate with endometrial adenocarcinomas. Interestingly, the regulation of human Hsp27 by estrogens and heat shock is reminiscent of the ecdysteroid and heat shock activation of the drosophila Hsp20 genes.
For IP: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: HSP25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Guinea Pig