Order Entry
ContactUsLinkComponent
Anti-INCENP Rabbit Polyclonal Antibody
Anti-INCENP Rabbit Polyclonal Antibody
Catalog #: 89359-240
Supplier:  Genetex
Anti-INCENP Rabbit Polyclonal Antibody
Catalog #: 89359-240
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Inner Centromere Protein Antigens 135/155Kda
  • Clonality:
    Polyclonal
  • Gene ID:
    3619
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    INCENP
  • Conjugation:
    Unconjugated
  • Size:
    100 μl
  • Western blot:
    Yes
  • Antigen synonyms:
    INCENP|MGC111393|FLJ31633
  • Storage buffer:
    Preservative: 15mM Sodium Azide; Constituents: 0.01M PBS. pH 7.4
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    13.9 mg/ml
  • Immunogen:
    Synthetic peptide: SKPRYHKRTSSAVWNSP (with N-terminal added cysteine) conjugated to KLH, corresponding to amino acids 884-901 of Human INCENP.
  • Tested applications:
    WB
  • Purification:
    IgG fraction
  • Cat. no.:
    89359-240

Specifications

About this item

Chromosomal passenger proteins are members of a group of proteins that move from centromeres to the spindle midzone during mitosis. INCENP (Inner Centromere Protein) is a chromosomal passenger protein with an essential role in mitosis and meiosis. It is a complex highly basic multidomain protein that is dynamically translocated from metaphase chromosomes to the spindle midzone during or just prior to anaphase. Some INCENP transfers to the equatorial cortex before formation of the cleavage furrow. Ultimately, it is discarded in the midbody at the completion of cytokinesis. INCENP, in complex with the passenger proteins aurora B kinase and survivin, acts at multiple points during mitosis. Studies in knockout mice established INCENP as essential for mouse development and viability. Loss of INCENP results in chromosome segregation defects and failures in cytokinesis. Targeting of survivin to its centromeric and central spindle locations during mitosis is dependent on INCENP. Direct binding of INCENP to beta tubulin has been demonstrated. Interactions of INCENP with heterochromatin protein HP1Hsa alpha and histone H2A.Z were also reported. INCENP levels are increased in several human cancer lines.

WB: Use at a minimum dilution of 1/2000 (recommended when using a whole extract of mouse NIH-3T3 and rat PC12 cells and a chemiluminescent detection reagent)- 1/5000 (recommended when using a nuclear extract of human HeLa cells and a chemiluminescent detection reagent). Predicted molecular weight: 106 kDa. Detection of the INCENP band by immunoblotting is specifically inhibited with the immunizing peptide. There is some debate about whether this antibody is suitable for use in IF and we would be most interested to hear from researchers who test this antibody in this application. The antibody has been reported to be suitable for use in IF at a dilution of 1/100 using HeLa cells. However, one of our customers used this product in HeLa cells and reported an unusual staining pattern - please see the enquiries section of the online datasheet for further details. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: INCENP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought