To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Histone H3
- Clonality:Monoclonal
- Clone:AH3-120
- Gene ID:3020
- Host:Mouse
- Isotype:IgG1
- Reactivity:Drosophila
- Antigen symbol:H3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:H3FA|HIST1H3J|H3FJ|HIST1H3E|HIST1H3G|HIST1H3B|H3FI|HIST1H3D|HIST1H3C|HIST1H3I|HIST1H3F|H3T|HIST3H3|H3FH|MGC126886|H3FD|H3FF|HIST1H3H|H3FC|H3FB|H3FT|H3FL|H3FK|HIST1H3A
- Modification:acetyl-Lys9
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: 0.01M PBS. pH 7.4
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:2.1 mg/ml
- Immunogen:A synthetic, acetylated histone H3 peptide (amino acids 7-20, acetyl K9) corresponding to the N-terminus of human histone H3, conjugated to KLH.
- Tested applications:ELISA, ICC/IF, IHC, WB
- Purification:Immunogen affinity purified
- Cat. no.:89359-236
Specifications
About this item
Histone proteins H3, H4, H2A, and H2B function as building blocks to package eukaryotic DNA into repeating nucleosome units that are folded in higher order chromatin fibers. The nucleosome is composed of an octamer containing a H3/H4 tetramer and two H2A/H2B dimers, surrounded by approximately 146 base pairs of DNA. A diverse and elaborate array of post-translational modifications including acetylation, phosphorylation, methylation, ubiquitination, and ADP-ribosylation occurs on the N-terminal tail domains of histones. Acetylation of lysine residues within these N-terminal domains by histone acetyl-transferases (HATs), including Gcn5p, P/CAF, p300/CBP, and TAFII250, is associated with transcriptional activation. This modification results in remodeling of the nucleosome structure into an open conformation more accessible to transcription complexes. Conversely, histone deacetylation by histone deacetylases (HDACs) is associated with transcription repression reversing the chromatin remodeling process. In most species, histone H3 is primarily acetylated at lysine 9, 14, 18, and 23. Acetylation at lysine 9 appears to have a dominant role in histone deposition and chromatin assembly in some organisms.
WB: Use at a concentration of 1 - 2 ?g/ml. Detects a band of approximately 17 kDa. (Determined using a whole cell extract of mouse fibroblasts 3T3 cell line treated with sodium butyrate.)Indirect ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher. We advise you to centrifuge this product vial before use.
Type: Primary
Antigen: H3F3A
Clonality: Monoclonal
Clone: AH3-120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Drosophila, Rat