To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Histone Deacetylase 2
- Clonality:Monoclonal
- Clone:HDAC2-62
- Gene ID:3066
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Bovine
- Antigen symbol:HDAC2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:HD2|YAF1|RPD3
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: 0.01M PBS. pH 7.4
- Storage temperature:Aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles.
- Concentration:2.1 mg/ml
- Immunogen:Synthetic peptide: SGEKTDTKGTKSEQLSNP with N-terminally added Cysteine conjugated to KLH, corresponding to amino acids 471-488 of Human HDAC2.
- Tested applications:ELISA, IHC, IP, WB
- Purification:Protein A purified
- Cat. no.:89359-226
Specifications
About this item
The basic repeating unit of chromatin is the nucleosome, which is composed of a protein octamer containing two each of the core histones H2A, H2B, H3, and H4, surrounded by approximately 146 base pairs of DNA. Reversible acetylation of highly conserved lysine residues in the N-terminal tail domains of core histones plays an important role in transcriptional regulation, cell cycle progression, and development events. Several histone acetyltransferases (HATs) catalyze this acetylation reaction (e.g. GCN5, PCAF, p300/CBP, TAFII250, P/CAF, SRC-1, BRCA-2). Acetylation of the core histones is generally considered to be associated with gene activation, probably through maintenance of the unfolded structure of transcribing nucleosomes. Histone acetylation is a dynamic process in which levels are determined by the net activities of HATs and the competing enzymes histone deacetylases (HDACs). Both activities are associated with the nuclear matrix. Eleven different mammalian HDACs have been described. HDACs 1-3 & 8 (Class I) are similar to yeast Rpd3 protein, while HDACs 4-7, 9 & 10 (Class II) are similar to yeast Hda1 protein. The activities of the histone deacetylases are often, but not always, associated with transcriptional repression and nucleosome condensation. HDAC1, HDAC2 and several others are the catalytic subunits of different multiprotein regulatory complexes. Other components of such complexes may include: corepressors such as mSin3, N-CoR, SMRT, associated proteins such as SAP18, SAP30, RbAp46, RbAp48, and c-Ski oncogenic protein (involved in DNA methylation). Nucleosome remodeling and deacetylation (NRD) complexes containing HDAC1, HDAC2, Mi-2 (CH3, CH4) dermatomyositis specific autoantigen, and MAT2 (metastasis-associated protein) (related to MAT1) have been described. It is therefore assumed that ATP-dependent nucleosome remodeling activity and histone deacetylation may be interconnected or interdependent. Recruitment of the multiprotein complexes to promoter sites occurs by many sequence specific DNA-binding proteins such as unliganded nuclear hormone receptors, DP1-E2F, YY1, and Rb family of transcription factors, transcriptional repressors, and tumor suppressors (e.g. BRCA1). Aberrant recruitment of HDACs by various oncoproteins may occur in certain neoplastic diseases. It has been found that inhibition ofHDAC2 activity by valporic acid induces proteosomal degradation of HDAC2.
ELISA: Use at an assay dependent dilution. IHC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a concentration of 0.25 - 0.5 ?g/ml. Detects a band of approximately 55 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: HDAC2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine