To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Meiotic Recombination 11
- ChIP:Yes
- Clonality:Polyclonal
- Gene ID:855264
- Host:Rabbit
- Isotype:IgG
- Reactivity:S. cerevisiae,C. elegans
- Antigen symbol:MRE11
- Conjugation:Unconjugated
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:ATLD|HNGS1|MRE11 homolog A|meiotic recombination 11 homolog A|MRE11B|MRE11|meiotic recombination 11 homolog 1|double-strand break repair protein MRE11A|DNA recombination and repair protein|AT-like disease|MRE11 homolog 1|endo/exonuclease Mre11
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 and 0.01% (w/v) Sodium Azide
- Storage temperature:Store vial at –20 °C prior to opening. Aliquot contents and freeze at –20 °C or below for extended storage Avoid cycles of freezing and thawing.
- Concentration:1.17 mg/ml
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 578-590 of Saccharomyces cerevisiae (baker's yeast) Mre11 protein.
- Tested applications:ChIP, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89359-210
Specifications
About this item
Mre11 is an 80 kDa protein that acts as a Mn2+ dependent double stranded DNA 3' to 5' exonuclease and a single stranded DNA endonuclease. It is part of a larger complex involved in the nucleolytic end-processing of double strand breaks and telomere maintenance.
Recommended dilutions: ChIP: Use at an assay dependent dilution. Not yet tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MRE11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: