To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Protein Disulfide Isomerase Family A, Member 6
- Clonality:Polyclonal
- Gene ID:10130
- Host:Rabbit
- Isotype:IgG
- Reactivity:Fish
- Antigen symbol:P5
- Conjugation:Unconjugated
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:TXNDC7|P5|ERP5
- Storage buffer:Antiserum containing 0.01% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide: DIDLSDVELDDLGKDEL, corresponding to amino acids 476-492 of Human P5.
- Tested applications:WB
- Purification:Unpurified
- Cat. no.:89357-994
Specifications
About this item
P5 is a novel protein that was originally identified in humans. Functionally, P5 has been shown to have peptide binding ability and chaperone activity specific to certain proteins. P5 shares several structural similarities to PDI, an important protein of the ER. The deduced amino acid sequence of P5 shows that it contains the ER retention signal KDEL. P5 also contains two CXXC-like motifs, a motif responsible for oxidoreductase activity. These two CXXC motifs are found on the N- and C-terminus of the protein, one at each end. Studies suggest that the N-terminal CXXC motif is more important than the second for isomerase activity. Contrary to most of the PDI-like proteins, P5 is not stress-inducible.
Recommended Starting Dilutions:WB: 1:10000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: P5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Fish