To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ATPase, Na+/K+ transporting, beta 1 polypeptide
- Clonality:Polyclonal
- Gene ID:481
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ATP1B1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:ATP1B
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:35 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.5mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human Sodium Potassium ATPase Beta 1. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89357-520
- Supplier No.:GTX113390
Specifications
About this item
Rabbit Polyclonal antibody to Sodium Potassium ATPase Beta 1 (ATPase, Na+/K+ transporting, beta 1 polypeptide)
Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: ATP1B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human