To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tubulin Epsilon 1
- Clonality:Monoclonal
- Clone:TUB-11
- Gene ID:51175
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:TUBE1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:TUBE|RP1-142L7.1|FLJ22589|FLJ44203|dJ142L7.2
- Amino acid number:352 to 366
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: 1% BSA, 0.01M PBS. pH 7.4
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:2.4mg/ml
- Immunogen:Synthetic peptide conjugated to KLH, corresponding to amino acids 352-366 of Human epsilon Tubulin.
- Tested applications:ELISA, WB
- Purification:Protein A purified
- Cat. no.:89357-384
- Supplier No.:GTX11322
Specifications
About this item
Mouse monoclonal [TUB - 11] to epsilon Tubulin
ELISA: Use at an assay dependent dilution. WB: Use at a concentration of 2 - 4 ?g/ml. Detects a band of approximately 55 kDa (predicted molecular weight: 52.9 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: TUBE1
Clonality: Monoclonal
Clone: TUB-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse