To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tubulin Beta 3
- Clonality:Monoclonal
- Clone:ONS.1A6
- Gene ID:10381
- Host:Mouse
- Isotype:IgG1
- Reactivity:Bovine
- Antigen symbol:TUBB3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 μl
- Western blot:Yes
- Format:Ascites
- Antigen synonyms:Tubulin beta-3 chain|CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: Ascites fluid.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide corresponding to the C-terminal sequence, coupled to BSA.
- Tested applications:ELISA, IHC, WB
- Purification:Ascites
- Cat. no.:89357-348
- Supplier No.:GTX11315
Specifications
About this item
Mouse monoclonal [ONS.1A6] to beta IV Tubulin
ELISA: Use at an assay dependent dilution. IHC: Use at an assay dependent dilution. Use with 10% neutral buffered formalinalcoholic formalin-xylene, paraffin-embedded , or 1-2% glutaraldehyde-4% formaldehyde fixed, LR Gold embedded. WB: Use at a dilution of 1/400. Detects a band of approximately 55 kDa and possibly the 100 kDa dimer. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: TUBB3
Clonality: Monoclonal
Clone: ONS.1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Hamster, Bovine