To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Pan Cytokeratin
- Clonality:Monoclonal
- Clone:C-11
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:p-CK
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µL
- Western blot:Yes
- Antigen synonyms:19|wide spectrum Cytokeratin|[cytokeratins 13|P-CK|24].|Cytokeratins|17|pan-cytokeratin|14|pan CK|16|pan-CK
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: 0.01M PBS (pH7.4)
- Storage temperature:–20 °C
- Immunogen:Keratin-enriched preparation from cultured human epidermoid carcinoma cell line A431.
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:purified by Protein A chromatography
- Cat. no.:89356-984
- Supplier No.:GTX11212
Specifications
About this item
GT211212 is a broad spectrum antibody which reacts specifically with a wide variety of normal, reactive and neoplastic epithelial tissues. The antibody reacts with simple, cornifying and non-cornifying squamous epithelia and pseudostratified epithelia. It does not react with non-epithelial normal human tissues. This antibody can be applied to methanol- or acetone-fixed frozen sections, and to formalin-fixed, paraffin-embedded human tissues. Increased staining intensity is seen following proteolytic treatment of tested tissue (protease iunmaskingi). Similarly embedded methacarn-fixed material is also suitable for cytokeratin demonstration. Anti-Pan Cytokeratin exhibits a wide interspecies cross-reactivity down to xenopus laevis (e.g., human, bovine, rat, and frog). GT211212 may be used for the localization of cytokeratins using various immunochemical assays including direct immunofluorescence using frozen or fixed tissue sections or cultured cells. It may also be used in flow cytometry of normal and neoplastic epithelial cells. Intermediate-sized filaments are abundant cytoplasmic structural proteins in most vertebrate cells.
Type: Primary
Antigen: p-CK
Clonality: Monoclonal
Clone: C-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human