To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-11A
- Clonality:Monoclonal
- Clone:2F3
- Gene ID:8766
- Host:Mouse
- Isotype:IgM
- Reactivity:Dog
- Antigen symbol:RAB11A
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:RAB11A|MGC1490|YL8|Ras-related protein Rab-11A|RAB 11A|member oncogene family|member RAS oncogene family
- Amino acid number:178 to 188
- Storage buffer:Phosphate-buffered saline
- Sequence:KQMSDRRENDM
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1.2mg/ml
- Immunogen:Rab 11a peptide KQMSDRRENDM, corresponding to amino acids 178-188 of Human Rab11A.
- Tested applications:ELISA, WB
- Purification:Purified IgM
- Cat. no.:89356-286
- Supplier No.:GTX11036
Specifications
About this item
Mouse monoclonal [2F3] to Rab11A
ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 26 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: RAB11A
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Dog