To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Hydroxysteroid 11-beta Dehydrogenase 2
- Clonality:Polyclonal
- Gene ID:3291
- Host:Rabbit
- Isotype:IgG
- Reactivity:Sheep
- Antigen symbol:HSD11B2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:HSD11K|AME|HSD2|SDR9C3|AME1
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:44 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human HSD11B2. The exact sequence is proprietary.
- Tested applications:Western Blot-Ag
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89356-054
- Supplier No.:GTX109786
Specifications
About this item
Rabbit polyclonal antibody to HSD11B2 (hydroxysteroid (11-beta) dehydrogenase 2)
IHC-Fr:Assay dependent.Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: HSD11B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep