To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast Growth Factor Receptor 3
- Clonality:Polyclonal
- Gene ID:2261
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:FGFR3
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:CEK2|HSFGFR3EX|CD333|JTK4|ACH
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: 1% BSA, 10mM Sodium Phosphate
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Synthetic peptide: AEEELVEADEAGSVK conjugated with glutaraldehyde to KLH, corresponding to amino acids 359-372 of Human FGFR3 with a C-terminal.
- Tested applications:IHC-P, IP, WB
- Purification:Immunogen affinity purified
- Cat. no.:89354-720
- Supplier No.:GTX10651
Specifications
About this item
Rabbit polyclonal to FGFR3
IHC-P: Use at a dilution of 1/1000 (determined by indirect immunoperoxidase staining of protease-digested, formalin-fixed, paraffin-embedded human and animal tissue sections). IP: Use at a dilution of 1/500.(determined by immunoprecipitation of FGFR3 using a whole cell lysate of 293T cells transfected with the human FGFR3 full length gene). WB: Use at a dilution of 1/500. Detects a band of approximately 110 and 120 kDa. (determined by western blotting using an extract of 293T cells transfected with the human FGFR-3 full length gene). The epitope(s) recognized by the antibody is resistant to routine formalin-fixation and paraffin-embedding. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: FGFR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human