To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast Growth Factor Receptor 3
- Clonality:Polyclonal
- Gene ID:2261
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:FGFR3
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:CEK2|HSFGFR3EX|CD333|JTK4|ACH
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: 1% BSA, 0.01M PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1.4mg/ml
- Immunogen:Synthetic peptide: KDLLPPAPPSSGGSRT, corresponding to amino acids 792-806 of the cytoplasmic region of Human FGFR3 with N-terminal added lysine. The peptide is conjugated to KLH with glutaraldehyde.
- Tested applications:IHC-P, IP, WB
- Purification:Immunogen affinity purified
- Cat. no.:89354-716
- Supplier No.:GTX10649
Specifications
About this item
Rabbit polyclonal to FGFR3
IHC-P: Use at a dilution of 1/1 - 1/250. IP: Use at a dilution of 1/2000 as determined by immunoprecipitation using a whole cell lysate of transfected 293T cells expressing recombinant human FGFR3. WB: Use at a dilution of 1/1000 as determined by western blotting using a whole cell extract of transfected 293T cells expressing recombinant human FGFR3. Detects a band of approximately 110-120 kDa. The epitope(s) recognized by the antibody is resistant to routine formalin-fixation and paraffin embedding. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: FGFR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human