To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast Growth Factor Receptor 2
- Clonality:Polyclonal
- Gene ID:2263
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:FGFR2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:TK14|ECT1|TK25|BEK|JWS|KGFR|CFD1|CEK3|CD332|K-SAM|BBDS|BFR-1
- Storage buffer:0.01M Phosphate-buffered saline, pH7.4, containing 1% bovine serum albumin and 15mM sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Synthetic peptide: KLPQYPHIGSVKT conjugated to KLH by a Glutaraldehyde linker, corresponding to amino acids 809-821 of the cytoplasmic region of Human FGFR2.
- Tested applications:IHC, IP, WB
- Purification:Immunogen affinity purified
- Cat. no.:89354-710
- Supplier No.:GTX10647
Specifications
About this item
Rabbit polyclonal to FGFR2
IHC: Use at an assay dependent dilution, this concentration is determined by indirect immunoperoxidase staining of trypsin or protease-digested, formalin-fixed, paraffin-embedded human and animal tissue sections. IP: Use at a dilution of 1:2,000. This concentration is determined by immunoprecipitation using a whole lysate of transfected cells expressing recombinant human FGFR2. WB: Use at a dilution of 1:2,000. This concentration is determined by immunoblotting using a whole extract of transfected cells expressing recombinant human FGFR2. Predicted molecular weight: 110 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: FGFR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat