To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cell division cycle 7-related protein kinase
- Clonality:Monoclonal
- Clone:DCS-341
- Gene ID:8317
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CDC7
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:CDC7L1|huCDC7|HsCDC7|Hsk1
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: 0.01M PBS. pH 7.4
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1.9mg/ml
- Immunogen:Full length protein (Human).
- Tested applications:ELISA, ICC/IF, IP, WB
- Purification:Protein G purified
- Cat. no.:89354-210
- Supplier No.:GTX10535
Specifications
About this item
Mouse monoclonal [DCS - 341] to Cdc7 Kinase
ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a concentration of 0.4 µg/ml. Detects a band of approximately 55 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CDC7
Clonality: Monoclonal
Clone: DCS-341
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: