To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 26, Subfamily B, Polypeptide 1
- Clonality:Polyclonal
- Gene ID:56603
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CYP26B1
- Conjugation:Unconjugated
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:Cytochrome P450 26B1|Retinoic acid-metabolizing cytochrome|RHFCA|P450 26A2|P450RAI2|P450 retinoic acid-inactivating 2|P450RAI-2|CYP26A2
- Storage buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:58 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at -20oC or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human Cytochrome P450 26B. The exact sequence is proprietary.
- Tested applications:WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89354-026
- Supplier No.:GTX104932
Specifications
About this item
Rabbit polyclonal antibody to Cytochrome P450 26B (cytochrome P450, family 26, subfamily B, polypeptide 1)
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: CYP26B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: