To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Hydroxysteroid 11-beta Dehydrogenase 1
- Clonality:Polyclonal
- Gene ID:3290
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:HSD11B1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:corticosteroid 11-beta-dehydrogenase isozyme 1|HSD11B1|11-beta-hydroxysteroid dehydrogenase 1|HSD11L|HDL|MGC13539|short chain dehydrogenase/reductase family 26C|OTTHUMP00000034650|hydroxysteroid (11-beta) dehydrogenase 1|SDR26C1|HSD11B|member 1|11-DH|HSD11|11-beta-HSD1
- Storage buffer:1XPBS, 40% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:32 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human HSD11B1. The exact sequence is proprietary.
- Tested applications:ICC/IF, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89350-418
- Supplier No.:GTX104626
Specifications
About this item
Rabbit Polyclonal antibody to HSD11B1 (hydroxysteroid (11-beta) dehydrogenase 1)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000.IHC-Fr:Assay dependent. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: HSD11B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human