To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Insulin Receptor
- Clonality:Polyclonal
- Gene ID:3643
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:INSR
- Conjugation:Unconjugated
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:IR|IR-A|HHF5|CD220|Kinase InsR|IR-B
- Modification:phospho-Tyr972
- Storage buffer:Preservative: 0.05% Sodium Azide; Constituents: 50% Glycerol, PBS, 1mg/ml BSA
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic phosphopeptide derived from the region of the human Insulin Receptor that contains tyrosine 972.
- Tested applications:WB
- Purification:Immunogen affinity purified
- Cat. no.:89361-696
- Supplier No.:GTX25678
Specifications
About this item
Rabbit polyclonal to Insulin Receptor ( phospho Y972 )
WB: 1/1000. Detects a band of approximately 110 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: INSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse