AdjusTable® Workstation, Stainless Steel Solid Top, Stainless Steel Base, Eagle MHC™
Supplier: Eagle Group
Certificates
About this item
16 gauge, Type 304 brushed stainless steel solid top.
- Stainless steel base with four 33" stainless posts
- One stainless wire shelf and stainless C-frame
Load capacity: 600 lbs.
Customers Who Bought This Also Bought