Stainless Steel Flat Top Worktable with Tubular Base, 16-Gauge Type 304, Eagle MHC™
Supplier: Eagle Group
Certificates
About this item
16 gauge type 304 stainless steel flat top.
- ¹³/₈" diameter legs and ⁵/₄" diameter crossbracing are heavy gauge stainless steel
- Rolled edge front and rear with ends turned under 90°
- 35.13 to 36.13" adjustable working height
Load capacity: 350 lbs.
Ordering information: Accessories available include drawers and casters.
Customers Who Bought This Also Bought