Stainless Steel Worktable with Backsplash and Undershelf, 14-Gauge Type 304, Eagle MHC™
Supplier: Eagle Group
Certificates
About this item
14 Gauge Type 304 stainless steel top with 4¹/₂" backsplash at rear of table.
- 35¹/₈" to 36¹/₈" adjustable working height
- Rolled edge front with ends turned under 90°
1⁵/₈" diameter legs and adjustable undershelf are heavy gauge stainless steel.
.
Ordering information: Accessories available include drawers and casters, sold separately.