Stainless Steel Cleanroom Tables, Perforated Top, Eagle MHC™
Supplier: Eagle Group
Certificates
About this item
14-gauge, Type 304 stainless steel top.
- Perforated top allows for 40% laminar flow
- Knocked down stainless steel C-frame
Brushed stainless top features ¾" holes on 1" centers.
Ordering information: Alcohol wiped and cleanroom bagged. Shipped knocked top reducing freight costs and potential for freight damage. Other base frame options available; contact your Avantor sales representative.
Customers Who Bought This Also Bought