Order Entry
ContactUsLinkComponent
Wood Laminate Storage Cabinet, Justrite®
Wood Laminate Storage Cabinet, Justrite®
Catalog #: 56610-486
Supplier:  Justrite
Nonmetallic Acid/Corrosive Storage Cabinet
Wood Laminate Storage Cabinet, Justrite®
Catalog #: 56610-486
Supplier:  Justrite

Specifications

  • Description:
    Storage cabinet, Wood Laminate
  • Cat. no.:
    56610-486
  • Capacity:
    36× 2½ L
  • Supplier No.:
    24140

Specifications

About this item

These wood laminate cabinets are suitable for highly corrosive liquids such as phenol, nitric acid and sulfuric acid.

  • Color: blue

Long lasting wood construction provides exceptional strength along with stainless handles and hinges to resist corrosion from harsh acids.

Customers Who Bought This Also Bought